missing translation for 'onlineSavingsMsg'
Learn More

Bisacodyl, Mikromol™

Product Code. 17912666
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17912666 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17912666 Supplier Mikromol Supplier No. MM0031.00
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Bisacodyl
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bisacodyl
CAS 603-50-9
Analyte or Component Names Bisacodyl
Shipping Condition Room Temperature
Molecular Formula C22 H19 N O4
Certifications/Compliance ISO 17034
Synonym 4,4'-(2-Pyridinylmethylene)bisphenol 1,1'-diacetate, 4,4'-(2-Pyridinylmethylene)bisphenol diacetate (ester), 4,4'-(2-Pyridylmethylene)diphenol diacetate (ester), 4,4'-(2-Pyridylmethylene)diphenol diacetate, Bicol, Bis(p-acetoxyphenyl)-2-pyridylmethane, Bisacodyl, Brocalax, Broxalax, Contralax, Correctol Caplets, Correctol Tablets, DAMP, Dulcolan, Dulcolax, Durolax, Endokolat, Eulaxan, Feen-a-Mint Tablets, Fenilaxan, Godalax, Hillcolax, Ivilax, LA96a, Laco, Laxadin, Laxanin N, Laxans, Laxine, Laxorex, NSC 614826, Neolax, Nigalax, Perilax, Prepacol, Pyrilax, SK-Bisacodyl, Sanvacual, Stadalax, Telemin, Theralax, Ulcol, Videx, Zetrax
InChI Formula InChI=1S/C22H19NO4/c1-15(24)26-19-10-6-17(7-11-19)22(21-5-3-4-14-23-21)18-8-12-20(13-9-18)27-16(2)25/h3-14,22H,1-2H3
SMILES CC(=O)Oc1ccc(cc1)C(c2ccc(OC(=O)C)cc2)c3ccccn3
IUPAC Name [4-[(4-acetyloxyphenyl)-pyridin-2-ylmethyl]phenyl] acetate
Molecular Weight (g/mol) 361.39
Quantity 500 mg
Formula Weight 361.1314 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.