missing translation for 'onlineSavingsMsg'
Learn More

Bisoprolol Fumarate 1.0 mg/ml in Methanol (as free base), LoGiCal

Product Code. 30459175
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 30459175

Brand: LoGiCal LGCAMP0460.0801

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Bisoprolol Fumarate 1.0 mg/ml in Methanol (as free base)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bisoprolol fumarate
CAS 104344-23-2
Analyte or Component Names Bisoprolol fumarate
Shipping Condition Room Temperature
Molecular Formula 2 C18 H31 N O4 . C4 H4 O4
Concentration 1 mg/mL
Certifications/Compliance ISO 17034
Synonym Bisoprolol Fumarate, Bisoprolol hemifumarate
InChI Formula InChI=1 S/2C18H31NO4.C4H4O4/c2*1-14(2)19-11-17(20)13-23-18-7-5-16(6-8-18)12-21-9-10-22-15(3)4;5-3(6)1-2-4(7)8/h2*5-8,14-15,17,19-20 H,9-13H2,1-4H3;1-2 H,(H,5,6)(H,7,8)/b;;2-1+
SMILES CC(C)NCC(O)COc1ccc(COCCOC(C)C)cc1.CC(C)NCC(O)COc2ccc(COCCOC(C)C)cc2.OC(=O)\C=C\C(=O)O
IUPAC Name (E)-but-2-enedioic acid;1-(propan-2-ylamino)-3-[4-(2-propan-2-yloxyethoxymethyl)phenoxy]propan-2-ol
Solvent or Matrix Methanol
Industry Type Forensics
Molecular Weight (g/mol) 766.9582
Quantity 1 mL
Formula Weight 766.4616 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.