missing translation for 'onlineSavingsMsg'
Learn More

Biuret, Mikromol

Product Code. 30458280
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30458280 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30458280 Supplier Mikromol Supplier No. MM1467.010025
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Biuret, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Biuret
CAS 108-19-0
Analyte or Component Names Biuret
Shipping Condition Room Temperature
Molecular Formula C2 H5 N3 O2
Certifications/Compliance ISO 9001
Synonym Imidodicarbonic diamide, Biuret (8CI), Allophanamide, Allophanic acid amide, Allophanimidic acid, Carbamylurea, Desmodeyn 75, Dicarbamylamine, HO 1, HO 1 (biuret), Isobiuret, N 75, NSC 8020, Urea, (aminocarbonyl)-, Ureidoformamide
InChI Formula InChI=1S/C2H5N3O2/c3-1(6)5-2(4)7/h(H5,3,4,5,6,7)
Solution Type Neat
SMILES NC(=O)NC(=O)N
IUPAC Name carbamoylurea
Molecular Weight (g/mol) 103.08
Quantity 25 mg
Formula Weight 103.0382 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.