missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
BLMH/Bleomycin Hydrolase Polyclonal antibody specifically detects BLMH/Bleomycin Hydrolase in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
Specifications
Specifications
| Antigen | BLMH/Bleomycin Hydrolase |
| Applications | Western Blot, ELISA, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunoprecipitation 1-4 uL/mg of lysate |
| Formulation | 0.2 um filtered solution in PBS |
| Gene Alias | BHBMHBLM hydrolase, bleomycin hydrolase, EC 3.4.22.40 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant Human BLMH/Bleomycin Hydrolase (Accession#: NP_000377.1; Ser 2-Glu 455). |
| Purification Method | Antigen and protein A Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?