missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BLNK Antibody (SR2143), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22546-50ul
This item is not returnable.
View return policy
Description
BLNK Monoclonal antibody specifically detects BLNK in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| BLNK | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| SR2143 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| AGM4, B cell linker protein, BASH, B-cell adapter containing a SH2 domain protein, B-cell adapter containing a Src homology 2 domain protein, B-cell linker, B-cell linker protein, BLNK-s, Cytoplasmic adapter protein, LY57, MGC111051, SLP-65BLNK-S, SLP65Ly57, Src homology 2 domain-containing leukocyte protein of 65 kDa | |
| A synthesized peptide derived from human BLNK (Uniprot #: Q8WV28) | |
| 50 μg | |
| Adaptive Immunity, Immunology, Phospho Specific, Signal Transduction | |
| 29760 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction