missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BLNK Antibody (SR2143), Novus Biologicals™
Rabbit Monoclonal Antibody
349.00 € - 449.00 €
Specifications
| Antigen | BLNK |
|---|---|
| Clone | SR2143 |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18681954
|
Novus Biologicals
NBP3-22546-100ul |
100 μg |
449.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18682665
|
Novus Biologicals
NBP3-22546-50ul |
50 μg |
349.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BLNK Monoclonal antibody specifically detects BLNK in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
| BLNK | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| AGM4, B cell linker protein, BASH, B-cell adapter containing a SH2 domain protein, B-cell adapter containing a Src homology 2 domain protein, B-cell linker, B-cell linker protein, BLNK-s, Cytoplasmic adapter protein, LY57, MGC111051, SLP-65BLNK-S, SLP65Ly57, Src homology 2 domain-containing leukocyte protein of 65 kDa | |
| A synthesized peptide derived from human BLNK (Uniprot #: Q8WV28) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR2143 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Adaptive Immunity, Immunology, Phospho Specific, Signal Transduction | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 29760 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title