missing translation for 'onlineSavingsMsg'
Learn More

Blue Tetrazolium chloride

Product Code. p-7020518
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11434144

Brand: Thermo Scientific Alfa Aesar A12502.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1871-22-3 | C40H36Cl2N8O2 | 731.68 g/mol

Blue Tetrazolium chloride is useful in the prepartion of DNA cation complexes and immunoadsorbents for anti-DNA antibodies. It is used in assay of reducing sugars. In combination with 4-nitrocatechol, tetrazolium blue chloride forms a ternary complex with molybdenum(VI) for extraction and spectrophotometric determination of molybdenum.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Blue Tetrazolium chloride is useful in the prepartion of DNA cation complexes and immunoadsorbents for anti-DNA antibodies. It is used in assay of reducing sugars. In combination with 4-nitrocatechol, tetrazolium blue chloride forms a ternary complex with molybdenum(VI) for extraction and spectrophotometric determination of molybdenum.

Solubility
Soluble in hot water (5 mg/ml), methanol (50 mg/ml), ethanol, and chloroform. Insoluble in acetone.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1871-22-3
Molecular Formula C40H36Cl2N8O2
Molecular Weight (g/mol) 731.68
MDL Number MFCD00040933
InChI Key RCEHREKDVGHYAM-UHFFFAOYSA-N
Synonym blue tetrazolium chloride, tetrazolium blue, tetrazolium blue chloride, blue tetrazolium, dimethoxy neotetrazolium, 4-anisyltetrazolium blue, 3,3'-3,3'-dimethoxy-1,1'-biphenyl-4,4'-diyl bis 2,5-diphenyl-2h-tetrazol-3-ium chloride, 3,3'-dianisole-4,4'-bis 3,5-diphenyltetrazolium chloride, 3,3'-dianisolebis 4,4'-3,5-diphenyl tetrazolium chloride, 3,3'-3,3'-dimethoxy-4,4'-biphenylene bis 2,5-diphenyl-2h-tetrazolium chloride
PubChem CID 9853362
ChEBI CHEBI:75198
IUPAC Name 2-[4-[4-(3,5-diphenyltetrazol-2-ium-2-yl)-3-methoxyphenyl]-2-methoxyphenyl]-3,5-diphenyltetrazol-2-ium;dichloride
SMILES [H+].[H+].[Cl-].[Cl-].COC1=CC(=CC=C1N1NC(=NN1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=C(N2NC(=NN2C2=CC=CC=C2)C2=CC=CC=C2)C(OC)=C1

Specifications

Melting Point ∼255°C (decomposition)
Quantity 1 g
Beilstein 3894077
Merck Index 14,9244
Solubility Information Soluble in hot water (5mg/ml),methanol (50mg/ml),ethanol,and chloroform. Insoluble in acetone.
Formula Weight 727.66
Chemical Name or Material Blue Tetrazolium chloride
compliance-icons
Product Identifier
  • Blue Tetrazolium chloride
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
Hazard Statement
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 2H-Tetrazolium,2,2'-(3,3'-dimethoxy[1,1'-biphenyl]-4,4'-diyl)bis[3,5-diphenyl-,dichloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.