missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Borrelia Burgdorferi Flagellar Antigen Antibody (7G11/6802) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-59982-0.025MG
This item is not returnable.
View return policy
Description
Borrelia Burgdorferi Flagellar Antigen Monoclonal specifically detects Borrelia Burgdorferi Flagellar Antigen in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Borrelia Burgdorferi Flagellar Antigen | |
| Monoclonal | |
| 1 mg/mL | |
| ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
| B burgdorferi, B. burgdorferi, B.burgdorferi, Lyme disease | |
| Native | |
| 0.025 mg | |
| Primary | |
| Bacteria | |
| Purified |
| Immunocytochemistry, ELISA, Immunofluorescence | |
| 7G11/6802 | |
| Unconjugated | |
| PBS (pH 7.2) with 0.09% Sodium Azide | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Reacts with a flagellar antigen. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction