missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Borrelia Burgdorferi Flagellar Antigen Antibody (7G11/6802) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
216.00 € - 511.00 €
Specifications
| Antigen | Borrelia Burgdorferi Flagellar Antigen |
|---|---|
| Clone | 7G11/6802 |
| Concentration | 1 mg/mL |
| Dilution | ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 |
| Applications | Immunocytochemistry, ELISA, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18276333
|
Novus Biologicals
NB110-59982-0.025MG |
0.025 mg |
216.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18705473
|
Novus Biologicals
NB110-59982 |
0.1 mg |
511.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Borrelia Burgdorferi Flagellar Antigen Monoclonal specifically detects Borrelia Burgdorferi Flagellar Antigen in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| Borrelia Burgdorferi Flagellar Antigen | |
| 1 mg/mL | |
| Immunocytochemistry, ELISA, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Bacteria | |
| Native | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 7G11/6802 | |
| ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| B burgdorferi, B. burgdorferi, B.burgdorferi, Lyme disease | |
| IgG2a | |
| Protein G purified | |
| Reacts with a flagellar antigen. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title