Learn More
BRE Mouse, anti-Human, CoraLite 594, Clone: 2E5A5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL594-66356-100UL
Description
The BRE antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human BRE. This antibody recognizes human, rat, mouse antigen. The BRE antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifications
| BRE | |
| 2E5A5 | |
| CoraLite 594 | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse | |
| 155 kDa | |
| IgG2a |
| Immunofluorescence | |
| 1000 μg/mL | |
| BRCA1 A complex subunit BRE, BRCC4, BRCC45, BRE | |
| 100 μL | |
| 9577 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |