missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRE Mouse, anti-Human, CoraLite 594, Clone: 2E5A5, Proteintech
Mouse Monoclonal Antibody
145.00 € - 450.00 €
Specifications
| Antigen | BRE |
|---|---|
| Clone | 2E5A5 |
| Concentration | 1000 μg/mL |
| Applications | Immunofluorescence |
| Conjugate | CoraLite 594 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18068415
|
Proteintech
CL594-66356-100UL |
100 μL |
450.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18034995
|
Proteintech
CL594-66356-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The BRE antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human BRE. This antibody recognizes human, rat, mouse antigen. The BRE antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifications
| BRE | |
| 1000 μg/mL | |
| CoraLite 594 | |
| Mouse | |
| 9577 | |
| IgG2a | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 2E5A5 | |
| Immunofluorescence | |
| Liquid | |
| Human, Rat, Mouse | |
| BRCA1 A complex subunit BRE, BRCC4, BRCC45, BRE | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title