missing translation for 'onlineSavingsMsg'
Learn More

Bromhexine Hydrochloride, LoGiCal

Product Code. 30458572
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30458572 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30458572 Supplier LoGiCal Supplier No. LGCFOR0065.00
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bromhexine hydrochloride
CAS 611-75-6
Analyte or Component Names Bromhexine hydrochloride
Shipping Condition Room Temperature
Molecular Formula C14 H20 Br2 N2 . Cl H
Certifications/Compliance ISO 9001
Synonym Benzenemethanamine, 2-amino-3,5-dibromo-N-cyclohexyl-N-methyl-, hydrochloride (1:1), Benzenemethanamine, 2-amino-3,5-dibromo-N-cyclohexyl-N-methyl-, monohydrochloride (9CI), 2-Amino-3,5-dibromo-N-cyclohexyl-N-methylbenzylamine monohydrochloride, Auxit, Bisolvon, Bisolvon hydrochloride, Bromhexine chloride, Bromhexine hydrochloride, Bromhexine monohydrochloride, Cyclohexylmethyl(2-amino-3,5-dibromobenzyl)ammonium chloride, N-Cyclohexyl-N-methyl-(2-amino-3,5-dibromobenzyl)ammonium chloride, N-Cyclohexyl-N-methyl-N-(2-amino-3,5-dibromobenzyl)ammonium chloride, Ophtosol, Quentan, Toluene-alpha,2-diamine, 3,5-dibromo-Nalpha-cyclohexyl-Nalpha-methyl-, monohydrochloride (8CI), Nalpha-Cyclohexyl-Nalpha-methyl-3,5-dibromotoluene-alpha,2-diamine hydrochloride
InChI Formula InChI=1S/C14H20Br2N2.ClH/c1-18(12-5-3-2-4-6-12)9-10-7-11(15)8-13(16)14(10)17;/h7-8,12H,2-6,9,17H2,1H3;1H
Solution Type Neat
SMILES Cl.CN(Cc1cc(Br)cc(Br)c1N)C2CCCCC2
IUPAC Name 2,4-dibromo-6-[[cyclohexyl(methyl)amino]methyl]aniline;hydrochloride
Industry Type Clinical Testing
Molecular Weight (g/mol) 412.59
Quantity 10 mg
Formula Weight 409.976 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.