missing translation for 'onlineSavingsMsg'
Learn More

alpha-Bromo-m-xylene, 96%

Product Code. 10182320 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 10182320

Brand: Thermo Scientific Acros 107560250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 620-13-3 | C8H9Br | 185.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 620-13-3
Molecular Formula C8H9Br
Molecular Weight (g/mol) 185.06
MDL Number MFCD00000177
InChI Key FWLWTILKTABGKQ-UHFFFAOYSA-N
Synonym 3-methylbenzyl bromide, 1-bromomethyl-3-methylbenzene, alpha-bromo-m-xylene, m-methylbenzyl bromide, m-xylyl bromide, benzene, 1-bromomethyl-3-methyl, 3-bromomethyl toluene, m-xylene, .alpha.-bromo, 3-methylbenzylbromide, m-xylene, alpha-bromo
PubChem CID 12099
IUPAC Name 1-(bromomethyl)-3-methylbenzene
SMILES CC1=CC(=CC=C1)CBr

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.3700g/mL
Boiling Point 185°C (340.0 mmHg)
Flash Point 82°C
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Linear Formula CH3C6H4CH2Br
Refractive Index 1.5640 to 1.568
Quantity 25 g
Beilstein 05, 374
Merck Index 15, 10288
Specific Gravity 1.37
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in alcohol and ether
Formula Weight 185.06
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material α-Bromo-m-xylene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • a-Bromo-m-xylene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.