missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BST2 Rabbit, anti-Human, Clone: 7J24, Proteintech
Rabbit Recombinant Monoclonal Antibody
149.00 € - 439.00 €
Specifications
| Antigen | BST2 |
|---|---|
| Clone | 7J24 |
| Concentration | 500 μg/mL |
| Applications | Western Blot, ELISA |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18099835
|
Proteintech
82733-7-RR-20UL |
20 μL |
149.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18060596
|
Proteintech
82733-7-RR-100UL |
100 μL |
439.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The BST2 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human BST2. This antibody recognizes human antigen. The BST2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| BST2 | |
| 500 μg/mL | |
| Liquid | |
| Human | |
| Bone marrow stromal antigen 2, BST 2, BST2, CD317, HM1.24 antigen, Tetherin | |
| Primary |
| 7J24 | |
| Western Blot, ELISA | |
| Rabbit | |
| 684 | |
| IgG | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title