missing translation for 'onlineSavingsMsg'
Learn More

BTAF1 Antibody [Alexa Fluor« 532], Novus Biologicals Biologicals™

Product Code. 30505780 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30505780 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30505780 Supplier Novus Biologicals Supplier No. NBP338188AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

BTAF1 Polyclonal antibody specifically detects BTAF1 in Human,Rat samples. It is validated for ChIP Assay,ELISA,Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen BTAF1
Applications ChIP Assay, ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias ATP-dependent helicase BTAF1, BTAF1 RNA polymerase II, B-TFIID transcription factor-associated, 170kDa (Mot1homolog, S. cerevisiae), B-TFIID transcription factor-associated 170 kDa subunit, EC 3.6.1, EC 3.6.4.-, MOT1MGC138406, TAF(II)170KIAA0940, TAF-172, TAF172BTAF1 RNA polymerase II, B-TFIID transcription factor-associated, 170 kD (Mot1homolog, S. cerevisiae), TAFII170TATA-binding protein-associated factor 172, TBP-associated factor 172
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1600-1849 of human BTAF1 (NP_003963.1).,, Sequence:, LAVQNSSLHDIQHAPKLSALKQLLLDCGLGNGSTSESGTESVVAQHRILIFCQLKSMLDIVEHDLLKPHLPSVTYLRLDGSIPPGQRHSIVSRFNNDPSIDVLLLTTHVGGLGLNLTGADTVVFVEHDWNPMRDLQAMDRAHRIGQKRVVNVYRLITRGTLEEKIMGLQKFKMNIANTVISQENSSLQSMGTDQLLDLFTLDKDGKAEKADTSTSGKASMKSILENLSDLWDQEQYDSEYSLENFMHSLK
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9044
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.