missing translation for 'onlineSavingsMsg'
Learn More

BTUM, TRC

Product Code. 30004130
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004130 10 g 10g
30007831 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30004130 Supplier Toronto Research Chemicals Supplier No. TRCB68936010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 151882-81-4
Molecular Formula C29 H28 N4 O6 S2
Molecular Weight (g/mol) 592.69
Synonym N,N'-[Methylenebis(4,1-phenyleneiminocarbonyl)]bis[4-methylbenzenesulfonamide], 4,4'-Bis(p-toluenesulfonylaminocarbonylamino)diphenylmethane, 4,4'-Bis(p-toluenesulfonylaminocarboxylamino)diphenylmethane, 4,4'-Bis(p-tolylsulfonylureido)diphenylmethane, BTUM
IUPAC Name 1-(4-methylphenyl)sulfonyl-3-[4-[[4-[(4-methylphenyl)sulfonylcarbamoylamino]phenyl]methyl]phenyl]urea
SMILES Cc1ccc(cc1)S(=O)(=O)NC(=O)Nc2ccc(Cc3ccc(NC(=O)NS(=O)(=O)c4ccc(C)cc4)cc3)cc2

Specifications

Quantity 10 g
InChI Formula InChI=1S/C29H28N4O6S2/c1-20-3-15-26(16-4-20)40(36,37)32-28(34)30-24-11-7-22(8-12-24)19-23-9-13-25(14-10-23)31-29(35)33-41(38,39)27-17-5-21(2)6-18-27/h3-18H,19H2,1-2H3,(H2,30,32,34)(H2,31,33,35)
Formula Weight 592.145
Chemical Name or Material BTUM
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.