missing translation for 'onlineSavingsMsg'
Learn More

Butoconazole nitrate, Thermo Scientific™

Product Code. 17585545 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17585545 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17585545 Supplier Thermo Scientific Acros Supplier No. 468770010
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Packaging Glass Bottle
CAS 64872-77-1
Certificate of Analysis Available
Physical Form Crystalline Powder
Molecular Formula C19H18Cl3N3O3S
MDL Number 00058159
Color White to Light Yellow
Melting Point 163°C
Merck Index 13-1525
InChI Key ZHPWRQIPPNZNML-UHFFFAOYNA-N
SMILES O[N+]([O-])=O.ClC1=CC=C(CCC(CN2C=CN=C2)SC2=C(Cl)C=CC=C2Cl)C=C1
IUPAC Name 1-[4-(4-chlorophenyl)-2-[(2,6-dichlorophenyl)sulfanyl]butyl]-1H-imidazole; nitric acid
Molecular Weight (g/mol) 474.78
Quantity 1 g
Chemical Name or Material Butoconazole nitrate
Show More Show Less
Product Title
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.