missing translation for 'onlineSavingsMsg'
Learn More

Butylhydroxytoluene, Mikromol™

Product Code. 17992856
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17992856 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17992856 Supplier Mikromol Supplier No. MM0679.000250
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Butylhydroxytoluene
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Butylhydroxytoluene
CAS 128-37-0
Analyte or Component Names Butylhydroxytoluene
Shipping Condition Room Temperature
Molecular Formula C15 H24 O
Certifications/Compliance ISO 17034
Synonym Butylhydroxytoluene, 2,6-Bis(1,1-dimethylethyl)-4-methylphenol, 2,6-Bis(tert-butyl)-4-methylphenol, 2,6-Di(tert-butyl)hydroxytoluene, 2,6-Di-tert-butyl-4-cresol, 2,6-Di-tert-butyl-4-hydroxytoluene, 2,6-Di-tert-butyl-4-methyl-1-hydroxybenzene, 2,6-Di-tert-butyl-4-methylhydroxybenzene, 2,6-Di-tert-butyl-4-methylphenol, 2,6-Di-tert-butyl-p-cresol, 2,6-Di-tert-butyl-p-cresole, 2,6-Di-tert-butyl-p-methylphenol, 2,6-Di-tert-butylcresol, 2,6-Di-tert-butylmethylphenol, 2,6-tert-Butyl-4-methylphenol, 3,5-Di-tert-butyl-4-hydroxytoluene, 4-Hydroxy-3,5-di-tert-butyltoluene, 4-Methyl-2,6-bis(1,1-dimethylethyl)phenol, 4-Methyl-2,6-di-tert-butylphenol, AO 29, AO 4, AO 4K, AOX 4, AOX 4K, Advastab 401, Agidol, Agidol 1, Agidol 1A, Alkofen BP, Antage BHT, Antioxidant 264, Antioxidant 29, Antioxidant 30, Antioxidant 4, Antioxidant 4K, Antioxidant DBPC, Antioxidant KB, Antioxidant MPJ, Antioxidant T 501, Antox QT, B-NOX BHT-P, BAT, BHT, BHT 264, BHT Swanox, BHT-C, Buks, Butylated hydroxytoluene, CAO 1, CAO 3, Catalin CAO 3, Chemanox 11, DBPC, Dalpac, Deenax, Di-tert-Butyl-4-methylphenol, Di-tert-butyl-p-cresol, Di-tert-butylcresol, Dibunol, Dibutylated hydroxytoluene, Dibutylcresol, Dibutylhydroxytoluene, E 321, Impruvol, Ionol, Ionol 1, Ionol 330, Ionol BHT, Ionol CP, Ionol K, Ionole, Kerabit, Lowinox BHT, Lubrizol 817, NSC 6347, Naugard BHT, Nocrac 200, Nocrac 2000, Nonox TBC, P 21, P 21 (phenol), Parabar 441, Permanax BHT, Popol, Ralox BHT, Selosol H 633, Stavox, Sumilizer BHT, Sumilizer BHT-R, Sustane BHT, Swanox BHT, T 501, T 501 (phenol), Tenamene 3, Tenox BHT, Tonarol, Topanol, Topanol BHT, Topanol O, Topanol O-FG, Topanol OC, Topanol OL, Ultranox 226, Vanlube PC, Vanlube PCX, Vanox PC, Vianol, Vulkanox KB, Yoshinox BHT, o-Di-tert-butyl-p-methylphenol, BHT, 2,6-di-tert-butyl-4-methylphenol
InChI Formula InChI=1S/C15H24O/c1-10-8-11(14(2,3)4)13(16)12(9-10)15(5,6)7/h8-9,16H,1-7H3
SMILES Cc1cc(c(O)c(c1)C(C)(C)C)C(C)(C)C
IUPAC Name 2,6-ditert-butyl-4-methylphenol
Molecular Weight (g/mol) 220.35
Quantity 250 mg
Formula Weight 220.1827 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.