missing translation for 'onlineSavingsMsg'
Learn More

Calreticulin Antibody, Novus Biologicals™

Product Code. p-5057396 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
Unit Size:
0.10mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18203822 0.025 mL 25µL
18218844 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18203822 Supplier Novus Biologicals Supplier No. NB600103SS
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 5 publications

Calreticulin Polyclonal specifically detects Calreticulin in Human, Mouse samples. It is validated for Gene Knock-Out, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), KnockDown, Simple Western, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calreticulin
Applications Gene Knock-Out, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), KnockDown, Simple Western, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:10000, Simple Western 1:500, Immunohistochemistry 1:20, Immunocytochemistry/Immunofluorescence 1:20-1:100. Use reported by customer review, Immunohistochemistry-Paraffin 1:20, Immunohistochemistry-Frozen reported in scientific literature (PMID 26643212), Knockout Validated, KnockDown Validated 1:2500
Formulation Whole antisera with 0.1% Sodium Azide
Gene Accession No. P14211
Gene Alias autoantigen Ro, calregulin, calreticulin, cC1qR, CRP55, CRT, CRTC, Endoplasmic reticulum resident protein 60, ERp60, FLJ26680, grp60, RO, Sicca syndrome antigen A (autoantigen Ro; calreticulin), SSAHACBP
Gene Symbols CALR
Host Species Rabbit
Immunogen Calreticulin Antibody was developed against a full-length protein to mouse Calreticulin [UniProt# P14211]
Molecular Weight of Antigen 48 kDa
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Autophagy, Cancer, Cellular Markers, ER Markers, Immunology, Membrane Trafficking and Chaperones, Neurodegeneration, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 811
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.