missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1QB Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99263-50ul
This item is not returnable.
View return policy
Description
C1QB Polyclonal antibody specifically detects C1QB in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| C1QB | |
| Polyclonal | |
| Immunohistochemistry 1:250-1:1000, Immunohistochemistry-Paraffin 1:250-1:1000 | |
| B chain, complement C1q subcomponent subunit B, complement component 1, q subcomponent, B chain, complement subcomponent C1q chain B, q subcomponent, beta polypeptide | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human C1QB | |
| 50 μg | |
| Alzheimers Research, Immunology, Innate Immunity, Neurodegeneration, Neuroscience | |
| 713 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction