missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1QB Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
227.00 € - 346.00 €
Specifications
| Antigen | C1QB |
|---|---|
| Dilution | Immunohistochemistry 1:250-1:1000, Immunohistochemistry-Paraffin 1:250-1:1000 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18607775
|
Novus Biologicals
NBP2-99263-100ul |
100 μg |
346.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18610136
|
Novus Biologicals
NBP2-99263-50ul |
50 μg |
227.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
C1QB Polyclonal antibody specifically detects C1QB in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| C1QB | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Alzheimers Research, Immunology, Innate Immunity, Neurodegeneration, Neuroscience | |
| 0.2 um filtered solution in PBS | |
| 713 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunohistochemistry 1:250-1:1000, Immunohistochemistry-Paraffin 1:250-1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse | |
| B chain, complement C1q subcomponent subunit B, complement component 1, q subcomponent, B chain, complement subcomponent C1q chain B, q subcomponent, beta polypeptide | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human C1QB | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title