missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CA125/MUC16 Antibody (X306cc), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NB120-10032-0.2MG
This item is not returnable.
View return policy
Description
CA125/MUC16 Monoclonal specifically detects CA125/MUC16 in Human samples. It is validated for Western Blot, ELISA, Microarray, Sandwich ELISA.
Specifications
| CA125/MUC16 | |
| Monoclonal | |
| Unconjugated | |
| Q8WXI7 | |
| MUC16 | |
| Full length native protein (purified from human ovarian carcinoma). | |
| 0.2 mg | |
| Extracellular Matrix | |
| 94025 | |
| Human | |
| IgG1 |
| Western Blot | |
| X306cc | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Microarray, Sandwich ELISA 1:100-1:2000 | |
| CA-125, CA125 ovarian cancer antigen, CA125MUC-16, FLJ14303, mucin 16, cell surface associated, mucin-16, Ovarian cancer-related tumor marker CA125, Ovarian carcinoma antigen CA125 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Epitope specificity group A (ISOBM classification). | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction