missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CA125/MUC16 Antibody (X306cc), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 3 publications
396.00 € - 559.00 €
Specifications
| Antigen | CA125/MUC16 |
|---|---|
| Clone | X306cc |
| Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Microarray, Sandwich ELISA 1:100-1:2000 |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18128989
|
Novus Biologicals
NB120-10032-0.2MG |
0.2 mg |
396.00 €
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18739143
|
Novus Biologicals
NB120-10032 |
0.5 mg |
559.00 €
0.50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CA125/MUC16 Monoclonal specifically detects CA125/MUC16 in Human samples. It is validated for Western Blot, ELISA, Microarray, Sandwich ELISA.Specifications
| CA125/MUC16 | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Microarray, Sandwich ELISA 1:100-1:2000 | |
| Unconjugated | |
| RUO | |
| Human | |
| CA-125, CA125 ovarian cancer antigen, CA125MUC-16, FLJ14303, mucin 16, cell surface associated, mucin-16, Ovarian cancer-related tumor marker CA125, Ovarian carcinoma antigen CA125 | |
| MUC16 | |
| IgG1 | |
| Protein A purified | |
| Epitope specificity group A (ISOBM classification). |
| X306cc | |
| Monoclonal | |
| Mouse | |
| Extracellular Matrix | |
| Q8WXI7 | |
| 94025 | |
| Full length native protein (purified from human ovarian carcinoma). | |
| Primary | |
| Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title