missing translation for 'onlineSavingsMsg'
Learn More

CACNA2D2 Antibody [FITC], Novus Biologicals Biologicals™

Product Code. 30488167 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30488167 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30488167 Supplier Novus Biologicals Supplier No. NBP335118F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CACNA2D2 Polyclonal antibody specifically detects CACNA2D2 in Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen CACNA2D2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias alpha 2 delta calcium channel subunit, CACNA2D, calcium channel, voltage-dependent, alpha 2/delta subunit 2, gene 26, KIAA0558Voltage-gated calcium channel subunit alpha-2/delta-2, LUAC11.1, voltage-dependent calcium channel subunit alpha-2/delta-2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 20-200 of human CACNA2D2 (NP_001167522.1).,, Sequence:, RPWPGCGPHPGPGTRRPTSGPPRPLWLLLPLLPLLAAPGASAYSFPQQHTMQHWARRLEQEVDGVMRIFGGVQQLREIYKDNRNLFEVQENEPQKLVEKVAGDIESLLDRKVQALKRLADAAENFQKAHRWQDNIKEEDIVYYDAKADAELDDPESEDVERGSKASTLRLDFIEDPNFKNK
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9254
Target Species Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.