missing translation for 'onlineSavingsMsg'
Learn More

CACYBP Antibody (7D11A8) [DyLight 550], Novus Biologicals Biologicals™

Product Code. 30487947 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30487947 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30487947 Supplier Novus Biologicals Supplier No. NBP327912R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CACYBP Monoclonal antibody specifically detects CACYBP in Human samples. It is validated for ELISA,Western Blot,Immunohistochemistry (Paraffin),Serum
TRUSTED_SUSTAINABILITY

Specifications

Antigen CACYBP
Applications ELISA, Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 7D11A8
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias CacyBP, calcyclin binding protein, GIG5, hCacyBP, MGC87971, RP1-102G20.6, S100A6-binding protein, S100A6BPgrowth-inhibiting gene 5 protein, Siah-interacting protein, Siah-interacting protein (SIP), SIPcalcyclin-binding protein
Host Species Mouse
Immunogen Truncated recombinant CACYBP expressed in E. Coli strain BL21 (DE3).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 27101
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.