missing translation for 'onlineSavingsMsg'
Learn More

Caffeine, Mikromol™

Product Code. 17910806
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17910806 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17910806 Supplier Mikromol Supplier No. MM0494.000250
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Caffeine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Caffeine
CAS 58-08-2
Analyte or Component Names Caffeine
Shipping Condition Room Temperature
Molecular Formula C8 H10 N4 O2
Synonym Caffeine, Theophylline-Ethylenediamine Imp. A (EP), Pentoxifylline Imp. F (EP), Dimenhydrinate Imp. C (EP), Theophylline Imp. A (EP), Theophylline-Ethylenediamine Anhydrous Imp. A (EP), Caffeine (8CI), 3,7-Dihydro-1,3,7-trimethyl-1H-purine-2,6-dione, 1,3,7-Trimethyl-2,6-dioxopurine, 1,3,7-Trimethyl-7H-purine-2,6-dione, 1,3,7-Trimethylxanthine, 7-Methyltheophylline, Alert-Pep, Asia migrine, Cafalgine, Cafeina, Caffedrine, Caffein, Cafipel, DHCplus, Dasin, Diurex, Durvitan, Guaranine, Koffein, Mateina, Methyltheobromine, Midron extra, Miudol, NSC 5036, New Cetamol, No-Doz, Palergot-C, Phensal, Refresh'n, Shape Plus, Stay Alert, Stim, Synalgos, Thein, Theine, Tri-Aqua, Vivarin, Wigraine, Dimenhydrinate Impurity C, Pentoxifylline Impurity F, Theophylline Impurity A, Theophylline-Ethylenediamine Impurity A
InChI Formula InChI=1S/C8H10N4O2/c1-10-4-9-6-5(10)7(13)12(3)8(14)11(6)2/h4H,1-3H3
SMILES CN1C(=O)N(C)c2ncn(C)c2C1=O
IUPAC Name 1,3,7-trimethylpurine-2,6-dione
Molecular Weight (g/mol) 194.19
Quantity 250 mg
Formula Weight 194.0804 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.