missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calnexin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-97476-0.2MG
This item is not returnable.
View return policy
Description
Calnexin Polyclonal specifically detects Calnexin in Human, Mouse, Rat, Porcine, Bovine, Canine, C. elegans, Chicken, Guinea Pig, Hamster, Primate, Rabbit, Sheep, Xenopus samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Single Cell Western.
Specifications
| Calnexin | |
| Polyclonal | |
| Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| calnexin, CNX, IP90FLJ26570, Major histocompatibility complex class I antigen-binding protein p88, P90 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 821 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide | |
| CANX | |
| Synthetic peptide corresponding to the sequence near the N-terminus of canine calnexin. | |
| 0.2 mg | |
| Primary | |
| Human, Mouse, Rat, Pig, Bovine, Canine, C. elegans, Chicken, Guinea Pig, Hamster, Primate, Rabbit, Sheep, Xenopus species | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction