missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calnexin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
180.00 € - 631.00 €
Specifications
| Antigen | Calnexin |
|---|---|
| Dilution | Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunofluorescence, Immunoprecipitation, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18084879
|
Novus Biologicals
NBP1-97476-0.025MG |
0.025 mg |
180.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18017509
|
Novus Biologicals
NBP1-97476-0.05MG |
0.05 mg |
415.00 €
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18067719
|
Novus Biologicals
NBP1-97476-0.2MG |
0.2 mg |
631.00 €
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Calnexin Polyclonal specifically detects Calnexin in Human, Mouse, Rat, Porcine, Bovine, Canine, C. elegans, Chicken, Guinea Pig, Hamster, Primate, Rabbit, Sheep, Xenopus samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Single Cell Western.Specifications
| Calnexin | |
| Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunofluorescence, Immunoprecipitation, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Rabbit, Sheep, Bovine, Canine, Hamster, Pig, Primate, Rat, Xenopus species, Mouse, C. elegans, Human, Guinea Pig, Chicken | |
| calnexin, CNX, IP90FLJ26570, Major histocompatibility complex class I antigen-binding protein p88, P90 | |
| CANX | |
| IgG | |
| Protein A purified |
| Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide | |
| 821 | |
| Synthetic peptide corresponding to the sequence near the N-terminus of canine calnexin. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title