missing translation for 'onlineSavingsMsg'
Learn More

Calpain 10 Antibody [CoraFluorÖ 1], Novus Biologicals Biologicals™

Product Code. 30517668 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30517668 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30517668 Supplier Novus Biologicals Supplier No. NBP338623CL1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Calpain 10 Polyclonal antibody specifically detects Calpain 10 in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calpain 10
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate CoraFluor 1
Formulation PBS
Gene Alias Calcium-activated neutral proteinase 10, calpain 10, calpain-10, calpain-like protease CAPN10, CANP 10, CANP10, EC 3.4.22.-, KIAA1845, NIDDM1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-230 of human Calpain 10 (NP_075571.1).,, Sequence:, MRAGRGATPARELFRDAAFPAADSSLFCDLSTPLAQFREDITWRRPQEICATPRLFPDDPREGQVKQGLLGDCWFLCACAALQKSRHLLDQVIPPGQPSWADQEYRGSFTCRIWQFGRWVEVTTDDRLPCLAGRLCFSRCQREDVFWLPLLEKVYAKVHGSYEHLWAGQVADALVDLTGGLAERWNLKGVAGSGGQQDRPGRWEHRTCRQLLHLKDQCLISCCVLSPRAG
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11132
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.