missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Calpastatin Polyclonal antibody specifically detects Calpastatin in Human, Mouse, Rat samples. It is validated for Western Blot, Gene Knock-Out
Specifications
Specifications
| Antigen | Calpastatin |
| Applications | Western Blot, Gene Knock-Out |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 - 1:2000, Knockout Validated |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | BS-17, Calpain inhibitor, calpastatin, MGC9402, Sperm BS-17 component |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to a sequence within amino acids 510-610 of human Calpastatin (NP_001177371.1). QNASSLKFEDAKLAAAISEVVSQTPASTTQAGAPPRDTSQSDKDLDDALDKLSDSLGQRQPDPDENKPMEDKVKEKAKAEHRDKLGERDDTIPPEYRHLLD |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?