missing translation for 'onlineSavingsMsg'
Learn More

Calretinin Antibody (CL12842) [Alexa Fluor« 594], Novus Biologicals Biologicals™

Product Code. 30527790 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30527790 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30527790 Supplier Novus Biologicals Supplier No. NBP344167AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Calretinin Monoclonal antibody specifically detects Calretinin in Human,Mouse samples. It is validated for Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calretinin
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL12842
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias 29 kDa calbindin, CAB29, CAL2, calbindin 2, calbindin 2, (29kD, calretinin), calbindin 2, 29kDa (calretinin), calbindin D29K, calretinin, CR
Host Species Mouse
Immunogen This antibody was generated using a recombinant protein sequence of P22676, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 794
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.