Learn More
CAMK2G Mouse, anti-Human, Clone: 1C12H10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68519-1-IG-150UL
Description
The CAMK2G antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CAMK2G. This antibody recognizes human, mouse, rat, pig, rabbit antigen. The CAMK2G antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| CAMK2G | |
| 1C12H10 | |
| CaM kinase II subunit gamma, CAMK, CAMK II, CaMK II subunit gamma, CAMK2G, CAMKG, FLJ16043 | |
| 150 μL | |
| 818 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Pig, Rabbit | |
| 150 kDa | |
| IgG1 |