missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CAMK2G Mouse, anti-Human, Clone: 1C12H10, Proteintech
Mouse Monoclonal Antibody
145.00 € - 410.00 €
Specifications
| Antigen | CAMK2G |
|---|---|
| Clone | 1C12H10 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, ELISA |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18097045
|
Proteintech
68519-1-IG-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18084265
|
Proteintech
68519-1-IG-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CAMK2G antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CAMK2G. This antibody recognizes human, mouse, rat, pig, rabbit antigen. The CAMK2G antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| CAMK2G | |
| 1000 μg/mL | |
| Liquid | |
| Human, Mouse, Rat, Pig, Rabbit | |
| CaM kinase II subunit gamma, CAMK, CAMK II, CaMK II subunit gamma, CAMK2G, CAMKG, FLJ16043 | |
| Primary |
| 1C12H10 | |
| Western Blot, ELISA | |
| Mouse | |
| 818 | |
| IgG1 | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title