Learn More
Candesartan, 98%, Thermo Scientific™
Angiotensin II receptor antagonist
66.20 € - 288.00 €
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15405129
|
Thermo Scientific Alfa Aesar
J62818.MC |
66.20 €
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
|
15495119
|
Thermo Scientific Alfa Aesar
J62818.03 |
1g |
288.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
SolubilitySoluble in ethyl acetate, methanol, water (<1mg/mL at 25°C), DMSO (88 mg/ml at 25°C), and ethanol (1 mg/ml at 25°C).
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Safety and Handling
- Candesartan
Signal Word
- Warning
Hazard Category
- Reproductive toxicity Category 2
Hazard Statement
- H361d-Suspected of damaging the unborn child.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contain: Candesartan
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.