missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Candesartan Cilexetil, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Candesartan Cilexetil
Specifications
Specifications
| Chemical Name or Material | Candesartan cilexetil |
| CAS | 145040-37-5 |
| Analyte or Component Names | Candesartan cilexetil |
| Shipping Condition | Room Temperature |
| Molecular Formula | C33 H34 N6 O6 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Candesartan Cilexetil, 2-Ethoxy-1-[[2'-(2H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-1H-benzimidazole-7-carboxylic acid 1-[[(cyclohexyloxy)carbonyl]oxy]ethyl ester, 1-[[(Cyclohexyloxy)carbonyl]oxy]ethyl 2-ethoxy-1-[[2'-(1H-tetrazol-5-yl)biphenyl-4-yl]methyl]benzimidazole-7-carboxylate, 2-Ethoxy-1-[(2'-(1H-tetrazol-5-yl)-1,1'-biphenyl-4-yl)methyl]-7-benzimidazolecarboxylic acid 1-(cyclohexyloxycarbonyloxy)ethyl ester, Atacand, Blopress, Candesartan cilexetil, TCV 116, TCY 116, (+/-)-2-Ethoxy-1-[[2'-(2H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-1H-benzimidazole-7-carboxylic acid 1-[[(cyclohexyloxy)carbonyl]oxy]ethyl ester, (+/-)-1-Hydroxyethyl 2-ethoxy-1-[p-(o-1H-tetrazol-5-ylphenyl)l]-7-benzimidazolecarboxylate, cyclohexyl carbonate (ester) |
| InChI Formula | InChI=1S/C33H34N6O6/c1-3-42-32-34-28-15-9-14-27(31(40)43-21(2)44-33(41)45-24-10-5-4-6-11-24)29(28)39(32)20-22-16-18-23(19-17-22)25-12-7-8-13-26(25)30-35-37-38-36-30/h7-9,12-19,21,24H,3-6,10-11,20H2,1-2H3,(H,35,36,37,38) |
| SMILES | CCOc1nc2cccc(C(=O)OC(C)OC(=O)OC3CCCCC3)c2n1Cc4ccc(cc4)c5ccccc5c6nnn[nH]6 |
| IUPAC Name | 1-cyclohexyloxycarbonyloxyethyl 2-ethoxy-3-[[4-[2-(1H-tetrazol-5-yl)phenyl]phenyl]methyl]benzimidazole-4-carboxylate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction