missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Canine/Feline SCF/c-kit Ligand Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB2268-SP
This item is not returnable.
View return policy
Description
SCF/c-kit Ligand Monoclonal specifically detects SCF/c-kit Ligand in Canine, Feline samples. It is validated for Western Blot.
Specifications
| SCF/c-kit Ligand | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q06220 | |
| KITLG | |
| E. coli-derived recombinant canine SCF/c-kit Ligand Lys26-Ala190 Accession # Q06220 | |
| 25 μg | |
| Primary | |
| Detects canine and feline SCF/c-kit Ligand in direct ELISAs and Western blots. In direct ELISAs and Western blots, 50%-100% cross-reactivity with recombinant human (rh) SCF and recombinant mouse SCF is observed and no cross-reactivity with rhM-CSF is observed. | |
| Canine, Feline | |
| Purified |
| Western Blot | |
| 371907 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| c-kit Ligand, DCUA, DFNA69, DKFZp686F2250, familial progressive hyperpigmentation 2, FPH2, FPHH, KIT ligand, Kitl, KITLG, KL-1, Mast cell growth factor, MGF, MGFSHEP7, SCFStem cell factor, SFc-Kit ligand, SHEP7, SLF, steel factor | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4254 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction