missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Canine/Feline SCF/c-kit Ligand Antibody, R&D Systems™

Mouse Monoclonal Antibody
196.00 € - 386.00 €
Specifications
| Antigen | SCF/c-kit Ligand |
|---|---|
| Clone | 371907 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18772749
![]() |
R&D Systems
MAB2268-SP |
25 μg |
196.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18182991
![]() |
R&D Systems
MAB2268 |
100 μg |
386.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SCF/c-kit Ligand Monoclonal specifically detects SCF/c-kit Ligand in Canine, Feline samples. It is validated for Western Blot.Specifications
| SCF/c-kit Ligand | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Canine, Feline | |
| Q06220 | |
| 4254 | |
| E. coli-derived recombinant canine SCF/c-kit Ligand Lys26-Ala190 Accession # Q06220 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects canine and feline SCF/c-kit Ligand in direct ELISAs and Western blots. In direct ELISAs and Western blots, 50%-100% cross-reactivity with recombinant human (rh) SCF and recombinant mouse SCF is observed and no cross-reactivity with rhM-CSF is observed. |
| 371907 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| c-kit Ligand, DCUA, DFNA69, DKFZp686F2250, familial progressive hyperpigmentation 2, FPH2, FPHH, KIT ligand, Kitl, KITLG, KL-1, Mast cell growth factor, MGF, MGFSHEP7, SCFStem cell factor, SFc-Kit ligand, SHEP7, SLF, steel factor | |
| KITLG | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title