missing translation for 'onlineSavingsMsg'
Learn More

Captopril Ethyl Ester, Mikromol™

Product Code. 30049151
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30049151 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30049151 Supplier Mikromol Supplier No. MM0067.03
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Captopril Ethyl Ester
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Captopril Ethyl Ester
CAS 86938-12-7
Analyte or Component Names Captopril Ethyl Ester
Shipping Condition Room Temperature
Molecular Formula C11 H19 N O3 S
Certifications/Compliance ISO 9001
Synonym 1-[(2S)-3-mercapto-2-methyl-1-oxopropyl]-L-Proline
InChI Formula InChI=1S/C11H19NO3S/c1-3-15-11(14)9-5-4-6-12(9)10(13)8(2)7-16/h8-9,16H,3-7H2,1-2H3/t8-,9+/m1/s1
SMILES CCOC(=O)[C@@H]1CCCN1C(=O)[C@H](C)CS
IUPAC Name ethyl (2S)-1-[(2S)-2-methyl-3-sulfanylpropanoyl]pyrrolidine-2-carboxylate
Molecular Weight (g/mol) 245.34
Quantity 100 mg
Formula Weight 245.1086 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.