missing translation for 'onlineSavingsMsg'
Learn More

CAPZA1 Antibody (2C8) [DyLight 488], Novus Biologicals Biologicals™

Product Code. 30518602 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30518602 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30518602 Supplier Novus Biologicals Supplier No. NBP342937G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CAPZA1 Monoclonal antibody specifically detects CAPZA1 in Human samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen CAPZA1
Applications ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone 2C8
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias Cap Z, CAPPA1, capping protein (actin filament) muscle Z-line, alpha 1, CAPZ, CapZ alpha-1, CAZ1, F-actin-capping protein subunit alpha-1
Host Species Mouse
Immunogen capping protein(actin filament) muscle Z-line, alpha 1 - (Uniprot# P52907)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 829
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.