missing translation for 'onlineSavingsMsg'
Learn More

Carbadox 100 μg/mL in Acetonitrile:Water, Dr. Ehrenstorfer

Product Code. 30314614
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30314614 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30314614 Supplier Dr. Ehrenstorfer Supplier No. DREA10968300WL100
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Carbadox
CAS 6804-07-5
Analyte or Component Names Carbadox
Shipping Condition Room Temperature
Molecular Formula C11 H10 N4 O4
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Carbazic acid, 3-(2-quinoxalinylmethylene)-, methyl ester, N,N'-dioxide (7CI), Carbazic acid, 3-(2-quinoxalinylmethylene)-, methyl ester, N1,N4-dioxide (8CI), Hydrazinecarboxylic acid, (2-quinoxalinylmethylene)-, methyl ester, N,N'-dioxide, Hydrazinecarboxylic acid, [(1,4-dioxido-2-quinoxalinyl)methylene]-, methyl ester (9CI), 2-Formylquinoxaline-1,4-dioxide carbomethoxyhydrazone, Carbadox, Fortigro, GS 6244, Getroxel, Mecadox, Methyl 3-(2-quinoxalinylmethylene)carbazate N1,N4-dioxide, Quinoxaline-2-carbomethoxyhydrazone 1,4-dioxide
InChI Formula InChI=1S/C11H10N4O4/c1-19-11(16)13-12-6-8-7-14(17)9-4-2-3-5-10(9)15(8)18/h2-7H,1H3,(H,13,16)/b12-6+
SMILES COC(=O)N\N=C\c1c[n+]([O-])c2ccccc2[n+]1[O-]
IUPAC Name methyl N-[(E)-(1,4-dioxidoquinoxaline-1,4-diium-2-yl)methylideneamino]carbamate
Solvent or Matrix Acetonitrile/Water
Molecular Weight (g/mol) 262.22
Quantity 1 mL
Formula Weight 262.0702 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.