missing translation for 'onlineSavingsMsg'
Learn More

Carbocisteine, Mikromol™

Product Code. 17997484
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17997484 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17997484 Supplier Mikromol Supplier No. MM0236.00
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Carbocisteine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Carbocisteine
CAS 638-23-3
Analyte or Component Names Carbocisteine
Shipping Condition Room Temperature
Molecular Formula C5 H9 N O4 S
Certifications/Compliance ISO 17034
Synonym Carbocisteine, Alanine, 3-[(carboxymethyl)thio]-, L- (6CI,8CI), (L)-2-Amino-3-(carboxymethylthio)propionic acid, (R)-S-(Carboxymethyl)cysteine, 3-[(Carboxymethyl)thio]-L-alanine, AHR 3053, Bronchokod, Carbocit, Carbocysteine, Chilvax, DF 1794Y, L-(Carboxymethyl)cysteine, LJ 206, Lisil, Lisomucil, Loviscol, Muciclar, Mucocis, Mucodyne, Mucofan, Mucolase, Mucolex, Mucopront, Mucotab, Mukinyl, NSC 14156, Pectox, Pulmoclase, Reomucil, Rhinathiol, Rhinatiol, Rinatiol, S-(Carboxymethyl)-(R)-cysteine, S-(Carboxymethyl)-L-cysteine, S-Carboxylmethyl-L-cysteine, Siroxyl, Thiodril, Transbronchin
InChI Formula InChI=1S/C5H9NO4S/c6-3(5(9)10)1-11-2-4(7)8/h3H,1-2,6H2,(H,7,8)(H,9,10)/t3-/m0/s1
SMILES N[C@@H](CSCC(=O)O)C(=O)O
IUPAC Name (2R)-2-amino-3-(carboxymethylsulfanyl)propanoic acid
Molecular Weight (g/mol) 179.19
Quantity 500 mg
Formula Weight 179.0252 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.