missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Carbonic Anhydrase VIII/CA8 Antibody (CA8/6814), Novus Biologicals™

Product Code. p-200086125 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30043242 100 μg 100µg
30043258 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30043242 Supplier Novus Biologicals™ Supplier No. NBP326747100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Carbonic Anhydrase VIII/CA8 Monoclonal antibody specifically detects Carbonic Anhydrase VIII/CA8 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Carbonic Anhydrase VIII/CA8
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CA8/6814
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT
Formulation 10 mM PBS with 0.05% BSA
Gene Alias CALScarbonate dehydratase, CAMRQ3, carbonic anhydrase VIIIMGC99509, carbonic anhydrase-like sequence, carbonic anhydrase-related protein, CARPCA-related protein, CA-VIII, MGC120502
Host Species Mouse
Immunogen Recombinant full-length human Carbonic Anhydrase VIII/CA8 protein
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 767
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.