missing translation for 'onlineSavingsMsg'
Learn More

Carprofen 1000 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30316300
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30316300 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30316300 Supplier Dr. Ehrenstorfer Supplier No. DREA11045850AL1000
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Carprofen
CAS 53716-49-7
Analyte or Component Names Carprofen
Shipping Condition +20°C
Molecular Formula C15 H12 Cl N O2
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym Carprofen, (2RS)-2-(6-Chloro-9H-carbazol-2-yl)propanoic acid
InChI Formula InChI=1S/C15H12ClNO2/c1-8(15(18)19)9-2-4-11-12-7-10(16)3-5-13(12)17-14(11)6-9/h2-8,17H,1H3,(H,18,19)
SMILES CC(C(=O)O)c1ccc2c(c1)[nH]c3ccc(Cl)cc23
IUPAC Name 2-(6-chloro-9H-carbazol-2-yl)propanoic acid
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 273.71
Quantity 1 mL
Formula Weight 273.0557 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.