missing translation for 'onlineSavingsMsg'
Learn More

Carprofen USP Related Compound A, Mikromol™

Product Code. 17951126
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17951126 25 mg 25mg
17982086 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17951126 Supplier Mikromol Supplier No. MM1484.010025
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Carprofen USP Related Compound A
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Carbazole
CAS 86-74-8
Analyte or Component Names Carbazole
Shipping Condition Room Temperature
Molecular Formula C12 H9 N
Certifications/Compliance ISO 9001
Synonym 9H-Carbazole, 9-Azafluorene, Chlorophenesin carbamate, Dibenzo[b,d]pyrrole, Dibenzopyrrole, Diphenylenimine, NSC 3498, SKF 20091, Carprofen Related Compound A
InChI Formula InChI=1S/C12H9N/c1-3-7-11-9(5-1)10-6-2-4-8-12(10)13-11/h1-8,13H
SMILES c1ccc2c(c1)[nH]c3ccccc23
IUPAC Name 9H-carbazole
Molecular Weight (g/mol) 167.21
Quantity 25 mg
Formula Weight 167.0735 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.