missing translation for 'onlineSavingsMsg'
Learn More

CARS Antibody (CL2304) - Azide and BSA Free, Novus Biologicals™

Product Code. 30297179 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Product Code. Quantity unitSize
30297179 100 μL 100µL
1 options
This item is not returnable. View return policy

Product Code. 30297179

Brand: Novus Biologicals NBP344481

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CARS Monoclonal antibody specifically detects CARS in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen CARS
Applications Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry
Classification Monoclonal
Clone CL2304
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, Immunohistochemistry-Paraffin 1:5000 - 1:10000
Formulation Lyophilized in 5% Trehalose.
Gene Alias CARS1, CysRS, cysteine translase, cysteine tRNA ligase 1, cytoplasmic, Cysteine--tRNA ligase, cysteinyl-tRNA synthetase, cysteinyl-tRNA synthetase, cytoplasmic, EC 6.1.1, EC 6.1.1.16, MGC:11246
Host Species Mouse
Immunogen This antibody was generated using a recombinant protein sequence of A8MVQ3, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 833
Reconstitution Centrifuge the vial of lyophilized antibody at 12,000 x g for 20 seconds. Reconstitute by adding sterile, distilled water to achieve a final antibody concentration of 1mg/mL.
Target Species Human
Content And Storage Store at 4⪚C short term. Aliquot and store at -20⪚C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.