missing translation for 'onlineSavingsMsg'
Learn More

CASK Antibody [PerCP], Novus Biologicals Biologicals™

Product Code. 30492620 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30492620 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30492620 Supplier Novus Biologicals Supplier No. NBP338000PCP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CASK Polyclonal antibody specifically detects CASK in Human samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen CASK
Applications ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate PerCP
Formulation PBS
Gene Alias Calcium/calmodulin-dependent serine protein kinase, calcium/calmodulin-dependent serine protein kinase (MAGUK family), calcium/calmodulin-dependent serine protein kinase membrane-associatedguanylate kinase, CAMGUK, CMG, EC 2.7.11, EC 2.7.11.1, FGS4CAGH39, FLJ22219, hCASK, LIN2FLJ31914, MICPCH, peripheral plasma membrane protein CASK, Protein lin-2 homolog, TNRC8, trinucleotide repeat containing 8
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human CASK (NP_001119526.1).,, Sequence:, MADDDVLFEDVYELCEVIGKGPFSVVRRCINRETGQQFAVKIVDVAKFTSSPGLSTEDLKREASICHMLKHPHIVELLETYSSDGMLYMVFEFMDGADLCFEIVKRADAGFVYSEAVASHYMRQILEALRYCHDNNIIHRDVKPHCVLLASKENSAPVKLGGFGVAIQLGESGLVAGGRVGTPHFMAPEVVKREPYGKPVDVWGCGVILFILLSGCLPFYGTKERLFEGIIKGKYKMNPRQWSHISESAKDLVRRMLMLDPAERITVYEALNHPWLKERDRYAYKIHLPETVEQLRKFNA
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 8573
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.