missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase-6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-87684-25ul
This item is not returnable.
View return policy
Description
Caspase-6 Polyclonal antibody specifically detects Caspase-6 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Caspase-6 | |
| Polyclonal | |
| Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Apoptotic protease Mch-2, CASP-6, caspase 6, apoptosis-related cysteine peptidase, caspase 6, apoptosis-related cysteine protease, caspase-6, EC 3.4.22, MCH2EC 3.4.22.59 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: MSSASGLRRGHPAGGEENMTETDAFYKREMFDPAEKYKMDHRRRGIALIFNHERFFWHLTLPERRGTCADRDNLTRRF | |
| 25 μL | |
| Apoptosis, Cancer, Caspases | |
| 839 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction