missing translation for 'onlineSavingsMsg'
Learn More

Catechol (Pyrocatechol; Benzene-1,2-diol), LoGiCal

Product Code. 30457594
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30457594 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30457594 Supplier LoGiCal Supplier No. LGCFOR0814.04

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Spezifikation

Chemical Name or Material Catechol
CAS 120-80-9
Analyte or Component Names Catechol
Shipping Condition Room Temperature
Molecular Formula C6 H6 O2
Certifications/Compliance ISO 9001
Synonym Benzene-1,2-diol (Pyrocatechol), Guaiacol Imp. A (EP), Benzene-1,2-diol, Pyrocatechol, 1,2-Benzenediol, 1,2-Dihydroxybenzene, 1,2-Hydroxybenzene, 2-Hydroxyphenol, C.I. 76500, C.I. Oxidation Base 26, Catechol, Catechol (phenol), Durafur Developer C, Fouramine PCH, Fourrine 68, NSC 1573, Oxyphenic acid, Pelagol Grey C, Phthalhydroquinone, Phthalic alcohol, Pyrocatechin, Pyrocatechine, Z 1, Z 1 (inhibitor), o-Benzenediol, o-Dihydroxybenzene, o-Dioxybenzene, o-Hydroquinone, o-Hydroxyphenol, o-Phenylenediol, Guaiacol Impurity A
InChI Formula InChI=1S/C6H6O2/c7-5-3-1-2-4-6(5)8/h1-4,7-8H
Solution Type Neat
SMILES Oc1ccccc1O
IUPAC Name benzene-1,2-diol
Industry Type Clinical Testing
Molecular Weight (g/mol) 110.11
Quantity 10 mg
Formula Weight 110.0368 g/mol
Mehr anzeigen Weniger anzeigen
Name des Produkts
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.