missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL21/6Ckine Antibody (MM0146-2H26) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12103-0.025MG
This item is not returnable.
View return policy
Description
CCL21/6Ckine Monoclonal specifically detects CCL21/6Ckine in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| CCL21/6Ckine | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:500-1:1000, Immunohistochemistry-Paraffin 15 ug/ml | |
| O00585 | |
| CCL21 | |
| Human CCL21 | |
| 0.025 mg | |
| Cancer, Cell Cycle and Replication | |
| 6366 | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry (Paraffin) | |
| MM0146-2H26 | |
| Unconjugated | |
| A 0.2 um filtered solution in PBS with No Preservative | |
| 6CkineSmall-inducible cytokine A21, Beta-chemokine exodus-2, chemokine (C-C motif) ligand 21, CKb9, member 21, SCYA21MGC34555, secondary lymphoid tissue chemokine, SLCSecondary lymphoid-tissue chemokine | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction