missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL21/6Ckine Antibody (MM0146-2H26) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
220.00 € - 533.00 €
Spezifikation
| Antigen | CCL21/6Ckine |
|---|---|
| Clone | MM0146-2H26 |
| Concentration | LYOPH |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|
18083897
|
Novus Biologicals
NBP2-12103-0.025MG |
0.025 mg |
220.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18068066
|
Novus Biologicals
NBP2-12103 |
0.1 mg |
533.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
CCL21/6Ckine Monoclonal specifically detects CCL21/6Ckine in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Spezifikation
| CCL21/6Ckine | |
| LYOPH | |
| Unconjugated | |
| Mouse | |
| Cancer, Cell Cycle and Replication | |
| O00585 | |
| 6366 | |
| IgG1 | |
| Protein G purified |
| MM0146-2H26 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 6CkineSmall-inducible cytokine A21, Beta-chemokine exodus-2, chemokine (C-C motif) ligand 21, CKb9, member 21, SCYA21MGC34555, secondary lymphoid tissue chemokine, SLCSecondary lymphoid-tissue chemokine | |
| CCL21 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts